missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ XUB Series Digital Ultrasonic Baths

Product Code. 11450998
Change view
Click to view available options
Capacity (Metric):
4.5 L
9.5 L
12.5 L
17.5 L
25 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11450998 9.5 L Each
11470998 17.5 L Each
11480998 25 L Each
11460998 12.5 L Each
11440998 4.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11450998 Supplier Grant Instruments™ Supplier No. XUB10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The XUB range of digitally controlled benchtop ultrasonic baths offer consistent and reliable performance in a variety of environments. Incorporating Frequency LEAP technology to ensure uniform levels of ultrasonic activity throughout the fluid, these baths offer high performance giving an accurate and precise ultrasonic process.

  • Adjustable speed control from 500 to 3000 rpm
  • Handles up to 32 tubes in three different sizes/combinations
  • Additional VP-1 flat head for single tube mixing supplied with unit
  • Rubber suction pads hold tight to the work surface and prevent the unit from walking - they also absorb vibration and prevent its transmission to the workbench
  • Easy to operate - select either Continuous or Touch operation and turn the knob to adjust the speed
  • The 32-socket universal platform for three tubes sizes (16 x 1.5mL, 8 x 0.5mL, 8 x 0.2mL) and a tip for mixing individual tubes up to 15mL
  • Operating temperature range: + 4°C to 40°C
TRUSTED_SUSTAINABILITY

Specifications

Timer 0 to 99 min.
Voltage 230 V
Capacity (Metric) 9.5 L
Depth (Metric) 183 mm
Temperature (Metric) Range Ambient +5°C to 70°C
Operating Frequency 32 to 38 kHz
compliance-icons
Product Identifier
  • Grant M2 ultrasonic solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: C9XYLATE, QUATERNARY FATTY AMINE ETHOXYLATE, BENZENESULFONIC ACID
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.