missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ XTR Adsorbent

Product Code. 10032841 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 pack
1000 g
50 Pack
500 g
5000 g
Unit Size:
1000g
500g
5kg
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10032841 500 g 500g
10761324 1000 g 1000g
10377971 5000 g 5kg
12491498 50 Pack Pack of 50
12481498 100 pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10032841 Supplier Macherey-Nagel™ Supplier No. 730595.500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use Chromabond™ XTR with Kieselguhr phase for liquid-liquid extraction. Chromabond™ XTR Adsorbent provides options for Chromabond™ XTR columns.

Features

  • Base material coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite)
  • large pore size, high pore volume, constantly high batch-to-batch quality
  • pH working range 1–13

Application

  • Liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel
  • High water loadability without breakthrough of water during elution with organic solvents
  • Also suited for removing small amounts of water from solvents which are not miscible with water

Advantages

  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation no formation of emulsions
  • High recovery rates
  • Saving of time and solvents
  • Organic solutions need not to be dried after separation

Solvents applicable for elution

  • diethyl ether
  • tert-butyl methyl ether
  • ethyl acetate
  • n-hexane
  • cyclohexane
  • toluene
  • dichloromethane (methylene chloride)
  • trichloromethane (chloroform)
  • solvent mixtures:
  • trichloromethane – methanol (90:10, v/v)
  • trichloromethane – methanol (85:15, v/v)
  • diethyl ether – ethanol (90:10, v/v)
  • diethyl ether – ethanol (80:20, v/v)
  • dichloromethane – 2-propanol (90:10, v/v)
  • dichloromethane – 2-propanol (85:15, v/v)

Specifications

Quantity 500 g
Product Type XTR Adsorbent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.