missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
XTP4 Polyclonal specifically detects XTP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | XTP4 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q9BRT3 |
| Gene Alias | C35, chromosome 17 open reading frame 37, HBV XAg-transactivated protein 4, HBV X-transactivated gene 4 protein, MGC14832, ORB3, protein C17orf37, Protein C35, XTP4RDX12 |
| Gene Symbols | MIEN1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?