missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XTP3TPA Rabbit anti-Human, Clone: 74, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90294-50UL
This item is not returnable.
View return policy
Description
XTP3TPA Monoclonal specifically detects XTP3TPA in Human samples. It is validated for ELISA.
Specifications
| XTP3TPA | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79077 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 74 | |
| ELISA 1:5000-1:10000 | |
| dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A | |
| Recombinant Human XTP3TPA Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction