missing translation for 'onlineSavingsMsg'
Learn More

XRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16888223 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16888223 20 μL 20µL
16878223 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16888223 Supplier Proteintech Supplier No. 112671AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XRN2 gene encodes a 5′-3′ exonuclease that promotes transcription termination at cotranscriptional cleavage sites. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen XRN2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene XRN2
Gene Accession No. Q9DBR1, Q9H0D6
Gene Alias 5 3 exoribonuclease 2, 5′ 3′ exoribonuclease 2, DHM1 like protein, DHP protein, XRN2
Gene Symbols Xrn2
Host Species Rabbit
Immunogen XRN2 Fusion Protein Ag1788
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22803, 24128, 362229
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.