missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRN2 Monoclonal Antibody (9F7G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531955
This item is not returnable.
View return policy
Description
MA5-31955 has been tested in indirect ELISA.
XRN2 gene encodes a 5'-3' exonuclease that promotes transcription termination at cotranscriptional cleavage sites. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| XRN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9H0D6 | |
| Xrn2 | |
| Purified recombinant fragment of human XRN2 (AA: 398-547) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 9F7G11 | |
| Unconjugated | |
| Xrn2 | |
| 5'-3' exoribonuclease 2; Dhm1; DHM1-like protein; DHP protein; Protein Dhm1; Xrn2; XRN2b | |
| Mouse | |
| Protein G | |
| RUO | |
| 22803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction