missing translation for 'onlineSavingsMsg'
Learn More

XRN2 Antibody (OTI5F4) [DyLight 755], Novus Biologicals™

Product Code. 30689015 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30689015 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689015 Supplier Novus Biologicals Supplier No. NBP344953IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XRN2 Monoclonal antibody specifically detects XRN2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRN2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI5F4
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 5'-3' exoribonuclease 2, DHM1-like protein, Dhm1-like protein (mouse homolog), DHP protein, EC 3.1.13, EC 3.1.13.-
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 677-779 of human XRN2(NP_036387) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22803
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.