missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC6BP1 Antibody (OTI5F3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74897AF647
This item is not returnable.
View return policy
Description
XRCC6BP1 Monoclonal specifically detects XRCC6BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| XRCC6BP1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATP23 | |
| Full length recombinant protein of human XRCC6BP1 (NP_150592) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5F3 | |
| Western Blot, Immunohistochemistry | |
| EC 3.4.24, EC 3.4.24.-, Ku70-binding protein 3, KUB3Ku70 binding protein 3, MGC134817, MGC134818, mitochondrial inner membrane protease ATP23 homolog, XRCC6 binding protein 1, XRCC6-binding protein 1 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 91419.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction