missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC4 Antibody (OTI4H9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74896G
This item is not returnable.
View return policy
Description
XRCC4 Monoclonal specifically detects XRCC4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| XRCC4 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| XRCC4 | |
| Full-length protein expressed in 293T cell transfected with human XRCC4 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4H9 | |
| Western Blot, Immunohistochemistry | |
| complementing defective, repair in Chinese hamster, X-ray repair complementing defective repair in Chinese hamster cells 4, X-ray repair cross-complementing protein 4 | |
| Mouse | |
| Affinity Purified | |
| Cancer, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
| 7518.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction