missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18494-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RecA/Rad51-related protein family that participates in homologous recombination to maintain chromosome stability and repair DNA damage. This gene functionally complements Chinese hamster irs1SF, a repair-deficient mutant that exhibits hypersensitivity to a number of different DNA-damaging agents and is chromosomally unstable. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. Alternatively spliced transcript variants encoding the same protein have been identified.Specifications
| XRCC3 | |
| Polyclonal | |
| Unconjugated | |
| XRCC3 | |
| DNA repair protein XRCC3, XRCC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102551085, 74335, 7517 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43542, Q9CXE6 | |
| Xrcc3 | |
| XRCC3 Fusion Protein Ag13317 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction