missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Xpress Monoclonal Antibody

Product Code. 10337642
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10337642 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10337642 Supplier Invitrogen™ Supplier No. R91025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

NOTE: R910-25 was previously sold under product #46-0528. When re-ordering, please use #R910-25. R910-25 is designed to specifically interact with the Xpress™ leader peptide (-Asp-Leu-Tyr-Asp-Asp-Asp-Asp-Lys-). R910-25 is tested in Western blot against 100 ng of an E. coli expressed fusion protein containing the Xpress epitope. This product contains enough material for 25 Western blots.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Epitope tag detection with an unconjugated antibody requires the use of a secondary antibody that carries a reporter for detection. Unconjugated antibodies let you choose the detection method which best suits your needs. They are also useful for the detection of weak signals, since a secondary antibody can amplify the signal. Xpress sequence: DLYDDDDK (Asp-Leu-Tyr-Asp-Asp-Asp-Asp-Lys).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Xpress
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias DLYDDDDK; Xpress Tag
Host Species Mouse
Immunogen Xpress synthetic peptide Asp-Leu-Tyr-Asp-Asp-Asp-Asp-Lys.
Purification Method Purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.