missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPR1 Recombinant Rabbit Monoclonal Antibody (HL2631)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556922
This item is not returnable.
View return policy
Description
XPR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
Specifications
| XPR1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9UBH6, Q9Z0U0 | |
| Xpr1 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human XPR1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2631 | |
| Unconjugated | |
| Xpr1 | |
| IBGC6; Protein SYG1 homolog; receptor for MCF virus 1; Rmc1; Rmc-1; RP11-46A10.1; suppressor of yeast G>a> deletion; susceptibility to xenotropic virus; Sxv; Syg1; X3; xenotropic and polytropic murine leukemia virus receptor; xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog; XPR1; XR; X-receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19775, 9213 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction