missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94001-0.02ml
This item is not returnable.
View return policy
Description
XPR1 Polyclonal antibody specifically detects XPR1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| XPR1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ90308, Protein SYG1 homolog, SYG1xenotropic and polytropic retrovirus receptor, X3, Xenotropic and polytropic murine leukemia virus receptor X3, xenotropic and polytropic retrovirus receptor 1, XR, X-receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 464-631 of human XPR1 (NP_001129141.1). DLKMDWGLFDKNAGENTFLREEIVYPQKAYYYCAIIEDVILRFAWTIQISITSTTLLPHSGDIIATVFAPLEVFRRFVWNFFRLENEHLNNCGEFRAVRDISVAPLNADDQTLLEQMMDQDDGVRNRQKNRSWKYNQSISLRRPRLASQSKARDTKVLIEDTDDEANT | |
| 0.02 mL | |
| Signal Transduction | |
| 9213 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction