missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPO5 Recombinant Rabbit Monoclonal Antibody (JE40-91)
GREENER_CHOICE

Product Code. 17145493
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17145493 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17145493 Supplier Invitrogen™ Supplier No. MA534823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: 293, rat epididymis tissue, human lung cancer tissue, human thyroid gland tissue, human breast tissue, mouse testis tissue, K562.

Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE40-91
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene XPO5
Gene Accession No. Q9HAV4
Gene Alias 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5
Gene Symbols XPO5
Host Species Rabbit
Immunogen Recombinant protein within Human Exportin-5 aa 1-240.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57510
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.