missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPNPEP3 Monoclonal Antibody (OTI4B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525652
This item is not returnable.
View return policy
Description
XPNPEP3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
XPNPEP3, also known as X-Prolyl Aminopeptidase 3, is a gene located on chromosome 22q that encodes an enzyme involved in the processing of mitochondrial matrix proteins. The enzyme is classified under the M24B subfamily of metallopeptidases, which specifically remove the N-terminal amino acid from oligopeptides where the penultimate residue is proline. XPNPEP3 is implicated in conditions such as cystic kidney disease and participates in TNF-TNFR2 cellular signaling pathways. Its yeast and plant homologs, like Icp55, perform similar functions by stabilizing proteins after import into the mitochondrial matrix. Mutations in XPNPEP3 are associated with defects in ciliary structure and function, contributing to the phenotypic spectrum seen in nephronophthisis-related ciliopathies (NPHP-RCs). The crystal structure of XPNPEP3 has been resolved at 1.65 A resolution, revealing its interaction with inhibitors such as apstatin. Furthermore, XPNPEP3 is identified as a transcriptional target of the canonical Wnt/beta-catenin signaling pathway, with elevated transcript levels observed in tumors exhibiting activated Wnt signaling.
Specifications
| XPNPEP3 | |
| Monoclonal | |
| 0.86 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B5DEQ3, B7ZMP1, Q9NQH7 | |
| XPNPEP3 | |
| Full length human recombinant protein of XPNPEP3 produced in HEK293 cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4B8 | |
| Unconjugated | |
| XPNPEP3 | |
| Aminopeptidase P3; APP3; E430012M05Rik; ICP55; Intermediate Cleaving Peptidase 55; LOW QUALITY PROTEIN: xaa-Pro aminopeptidase 3; NPHPL1; probable Xaa-Pro aminopeptidase 3; putative Xaa-Pro aminopeptidase 3; Xaa-Pro aminopeptidase 3; xaa-Pro aminopeptidase 3; probable Xaa-Pro aminopeptidase 3; XPNPEP3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3; X-prolyl aminopeptidase 3, mitochondrial; zgc:76905; zgc:76905 protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 321003, 481237, 63929, 685823 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction