missing translation for 'onlineSavingsMsg'
Learn More

XPNPEP3 Antibody (OTI5G2) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30687324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687324 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687324 Supplier Novus Biologicals Supplier No. NBP344640MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XPNPEP3 Monoclonal antibody specifically detects XPNPEP3 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPNPEP3
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI5G2
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Aminopeptidase P3, APP3probable Xaa-Pro aminopeptidase 3, NPHPL1EC 3.4.11.9, X-Pro aminopeptidase 3, X-prolyl aminopeptidase (aminopeptidase P) 3, putative
Host Species Mouse
Immunogen Full length human recombinant protein of human XPNPEP3(NP_071381) produced in HEK293 cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 63929
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.