missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPNPEP1 Recombinant Rabbit Monoclonal Antibody (SAIC-07E-1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547892
This item is not returnable.
View return policy
Description
Specificity: Recognises human XPNPEP1.
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Specifications
| XPNPEP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9NQW7 | |
| Xpnpep1 | |
| Peptide TLSLDEVYLIDSGAQYK derived from Xaa-Pro aminopeptidase 1 conjugated to KLH. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
| SAIC-07E-1 | |
| Unconjugated | |
| Xpnpep1 | |
| Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7511 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction