missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPNPEP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519139
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, porcine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Specifications
| XPNPEP1 | |
| Polyclonal | |
| Unconjugated | |
| Xpnpep1 | |
| Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100152688, 170750, 170751, 7511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O54975, Q6P1B1, Q9NQW7 | |
| Xpnpep1 | |
| Synthetic peptide sequence (LIRETQPISKQH) corresponding to the C-terminus amino acids of XPNPEP1 (aa 612-623). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction