missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | XPG |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
XPG Polyclonal specifically detects XPG in Human samples. It is validated for Western Blot.Specifications
| XPG | |
| Polyclonal | |
| Rabbit | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR | |
| ERCC5 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P28715 | |
| 2073 | |
| IgG | |
| 133 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title