missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPG Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97984AF594
This item is not returnable.
View return policy
Description
XPG Polyclonal specifically detects XPG in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| XPG | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR | |
| Produced in rabbits immunized with E. coli-derived Human XPG fragment. | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2073 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction