missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPG Antibody (8H7/XPG) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50366-0.025ml
This item is not returnable.
View return policy
Description
XPG Monoclonal antibody specifically detects XPG in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications
| XPG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR | |
| Recombinant human XPG protein produced in baculovirus | |
| 0.025 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2073 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| 8H7/XPG | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction