missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544878
This item is not returnable.
View return policy
Description
Peptide sequence: FLLRLYRKTF EKDSKAEEVW MKFRKEDSSK RIRKSHKRPK DPQNKERAST Sequence homology: Dog: 83%; Horse: 85%; Human: 100%; Rat: 85%.
The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
Specifications
| XPF | |
| Polyclonal | |
| Unconjugated | |
| ERCC4 | |
| AI606920; DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC excision repair 4, endonuclease catalytic subunit; ERCC11; ERCC4; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; FANCQ; RAD1; RGD1560340; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F; XFEPS; XPF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2072 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92889 | |
| ERCC4 | |
| Synthetic peptide directed towards the middle region of human ERCC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction