missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Xpect™ RSV Test

Product Code. 13245679
Change view
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13245679 30 Tests 30 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13245679 Supplier Thermo Scientific™ Supplier No. R24601
  • ONLINE EXCLUSIVE PRICE
    £1125.55
    £1012.90 / 30 tests
    Save £112.65
    10% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect Respiratory Syncytial Virus (RSV) from nasopharyngeal specimens in neonatal and pediatric patients and get results in 15 minutes.

Optimize your workflow by rapidly detecting Respiratory Syncytial Virus antigen (RSV) directly from nasopharyngeal specimens in neonatal and pediatric patients using Thermo Scientific™ Xpect™ RSV Test. Combine excellent clinical performance with a simple, 2-step procedure that offers accurate results within 15 minutes.

Use this rapid, lateral flow immunoassay for the direct qualitative detection of Respiratory Syncytial Virus (RSV). Reduce secondary testing, decrease unnecessary antibiotic use and guide appropriate antiviral therapy.
  • Improve patient management with accurate, easy-to-read results in just 15 minutes
  • Eliminate workflow bottlenecks with less than one minute hands-on time and a simple, walk-away procedure
  • Stock up with 18-month shelf life at room-temperature storage
  • Offers flexible specimen storage, collection, and transport options

Xpect RSV uses antibody-coated color particles to bind with target antigen in the sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.

Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.

Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Description Xpect RSV Test
Detectable Analytes Respiratory Syncytial Virus Antigen
Sample Type Nasopharyngeal
Type Reagent
Includes Test devices, extraction buffer (2 x 10mL), disposable extraction tubes, disposable pipettes
Quantity 30 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.