missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Xpect™ Giardia Test

Product Code. 13285619
Change view
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13285619 20 Tests/Kit 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13285619 Supplier Thermo Scientific™ Supplier No. R2450020
  • ONLINE EXCLUSIVE PRICE
    £520.00
    £510.00 / 20 tests
    Save £10.00
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect Giardia-specific antigen from fresh, frozen, or preserved human fecal specimens.

Optimize your workflow by rapidly detecting Giardia-specific antigen from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Giardia Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.

Use this rapid, lateral flow immunoassay for the direct qualitative detection of Giardia-specific antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
  • Clearly visible, easy-to-read results in just 15 minutes
  • Less than one minute hands-on time
  • 95.8 to 97.9% sensitivity
  • Single and combination immunoassays available
  • No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Xpect Giardia uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.

Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.

Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Xpect Giardia Test
Detectable Analytes Giardia
Sample Type Fecal
Type Giardia Testing Kit
Includes Test devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes and dilution tubes
Quantity 20 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.