missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Xpect™ Giardia/Cryptosporidium Test
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
Description
Optimize your workflow by rapidly detecting Giardia- and Cryptosporidium-specific antigens from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Giardia/Cryptosporidium Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Giardia- and Cryptosporidium-specific antigens. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
Xpect Giardia/Cryptosporidium uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. A blue line will appear at the giardia test position if giardia antigen is present. If cryptosporidium antigen is present, a red line will appear at the cryptosporidium test position. These lines will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
- Clearly visible, easy-to-read results in just 15 minutes
- Less than one minute hands-on time
- 95.8 to 97.9% sensitivity
- Single and combination immunoassays available
- No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Xpect Giardia/Cryptosporidium uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. A blue line will appear at the giardia test position if giardia antigen is present. If cryptosporidium antigen is present, a red line will appear at the cryptosporidium test position. These lines will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Xpect Giardia/Cryptosporidium Test |
| CE Marker | Yes |
| Detectable Analytes | Giardia and Cryptosporidium Antigen |
| Includes | Test devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes, dilution tubes |
| Quantity | 20 Tests/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction