missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPE/DDB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98940-50ul
This item is not returnable.
View return policy
Description
XPE/DDB2 Polyclonal antibody specifically detects XPE/DDB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| XPE/DDB2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human XPE/DDB2. | |
| 50 μg | |
| DNA Repair, Nucleotide Excision Repair | |
| 1643 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction