missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPE/DDB2 Antibody (24E5), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | XPE/DDB2 |
|---|---|
| Clone | 24E5 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043496
|
Novus Biologicals
NBP3-26631-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043596
|
Novus Biologicals
NBP3-26631-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XPE/DDB2 Monoclonal antibody specifically detects XPE/DDB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| XPE/DDB2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein | |
| A synthesized peptide derived from Human XPE/DDB2 [UniProt Q92466] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 24E5 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Nucleotide Excision Repair | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1643 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title