missing translation for 'onlineSavingsMsg'
Learn More

XPD Rabbit anti-Human, Mouse, Rat, Clone: 6T9K6, Novus Biologicals™

Product Code. 18343066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343066 100 μg 100µL
18355065 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18343066 Supplier Novus Biologicals Supplier No. NBP315704100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

XPD Monoclonal antibody specifically detects XPD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPD
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6T9K6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Basic transcription factor 2 80 kDa subunit, BTF2 p80, COFS2, CXPD, DNA excision repair protein ERCC-2, DNA repair protein complementing XP-D cells, EC 3.6.1, EC 3.6.4.12, EM9TFIIH basal transcription factor complex 80 kDa subunit, excision repair cross-complementing rodent repair deficiency, complementationgroup 2, MAG, MGC102762, MGC126218, MGC126219, TFIIH 80 kDa subunit, TFIIH basal transcription factor complex helicase subunit, TFIIH basal transcription factor complex helicase XPD subunit, TTD, xeroderma pigmentosum complementary group D, Xeroderma pigmentosum group D-complementing protein, XPDC, XPDTFIIH p80
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of human XPD (P18074).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2068
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.