missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPD Antibody (S3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002068-M04
This item is not returnable.
View return policy
Description
XPD Monoclonal antibody specifically detects XPD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| XPD | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Basic transcription factor 2 80 kDa subunit, BTF2 p80, COFS2, CXPD, DNA excision repair protein ERCC-2, DNA repair protein complementing XP-D cells, EC 3.6.1, EC 3.6.4.12, EM9TFIIH basal transcription factor complex 80 kDa subunit, excision repair cross-complementing rodent repair deficiency, complementationgroup 2, MAG, MGC102762, MGC126218, MGC126219, TFIIH 80 kDa subunit, TFIIH basal transcription factor complex helicase subunit, TFIIH basal transcription factor complex helicase XPD subunit, TTD, xeroderma pigmentosum complementary group D, Xeroderma pigmentosum group D-complementing protein, XPDC, XPDTFIIH p80 | |
| ERCC2 (AAH08346, 1 a.a. ~ 405 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRELKRTLDAKGHGVLEMPSGTGKTVSLLALIMAYQRAYPLEVTKLIYCSRTVPEIEKVIEELRKLLNFYEKQEGEKLPFLGLALSSRKNLCIHPEVTPLRFGKDVDGKCHSLTASYVRAQYQHDTSLPHCRFYEEFDAHGREVPLPAGIYNLDDLKALGRRQGWCPYFLARYSILHANVVVYSYHYLLDPKIADLVSKELARKAVVVFDEAHNIDNVCIDSMSVNLTRRTLDRCQGNLETLQKTVLRIKETDEQRLRDEYRRLVEGLREASAARETDAHLANPVLPDEVLQEAVPGSIRTAEHFLGFLRRLLEYVKWRLRVQHVVQESPPAFLSGLAQRVCIQRKPLRFCAERLRSLLHTLEITDLADFSPLTLLANFATLVSTYAKGQAQHCGSSRNQKRSHP | |
| 0.1 mg | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2068 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry | |
| S3 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| BC008346 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction