missing translation for 'onlineSavingsMsg'
Learn More

XPD Antibody (OTI4B11) [PerCP], Novus Biologicals™

Product Code. 30689405 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689405 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689405 Supplier Novus Biologicals Supplier No. NBP344677PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XPD Monoclonal antibody specifically detects XPD in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPD
Applications Western Blot
Classification Monoclonal
Clone OTI4B11
Conjugate PerCP
Formulation PBS
Gene Alias Basic transcription factor 2 80 kDa subunit, BTF2 p80, COFS2, CXPD, DNA excision repair protein ERCC-2, DNA repair protein complementing XP-D cells, EC 3.6.1, EC 3.6.4.12, EM9TFIIH basal transcription factor complex 80 kDa subunit, excision repair cross-complementing rodent repair deficiency, complementationgroup 2, MAG, MGC102762, MGC126218, MGC126219, TFIIH 80 kDa subunit, TFIIH basal transcription factor complex helicase subunit, TFIIH basal transcription factor complex helicase XPD subunit, TTD, xeroderma pigmentosum complementary group D, Xeroderma pigmentosum group D-complementing protein, XPDC, XPDTFIIH p80
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 78-307 of human ERCC2 (NP_000391) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2068
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.