Learn More
Invitrogen™ XPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA3956
Description
PA3-956 detects XPC in human and mouse cells. PA3-956 has been successfully used in Western blotting and ICC/IF procedures. By Western blot it detects a ~125 kDa protein representing XPC. The PA3-956 immunogen is two synthetic peptides, individually conjugated to the C-Terminus and N-Terminus respectively of human XPC.
This gene encodes a component of the nucleotide excision repair (NER) pathway. There are multiple components involved in the NER pathway, including Xeroderma pigmentosum (XP) A-G and V, Cockayne syndrome (CS) A and B, and trichothiodystrophy (TTD) group A, etc. This component, XPC, plays an important role in the early steps of global genome NER, especially in damage recognition, open complex formation, and repair protein complex formation. Mutations in this gene or some other NER components result in Xeroderma pigmentosum, a rare autosomal recessive disorder characterized by increased sensitivity to sunlight with the development of carcinomas at an early age. Alternatively spliced transcript variants have been found for this gene.
Specifications
| XPC | |
| Polyclonal | |
| Unconjugated | |
| XPC | |
| DNA repair protein complementing XP-C cells; DNA repair protein complementing XP-C cells homolog; mutant xeroderma pigmentosum group C; NER; p125; RAD4; Xeroderma pigmentosum group C-complementing protein; Xeroderma pigmentosum group C-complementing protein homolog; xeroderma pigmentosum, complementation group C; XP3; XPC; XPC complex subunit, DNA damage recognition and repair factor; XPCC | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole Serum with 0.05% sodium azide | |
| P51612, Q01831 | |
| XPC | |
| Two synthetic peptides, individually conjugated to the C-Terminus and N-Terminus respectively of human XPC. | |
| RUO | |
| 22591, 7508 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.