missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPC Monoclonal Antibody (3.26)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123328
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa whole cell extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a component of the nucleotide excision repair (NER) pathway. There are multiple components involved in the NER pathway, including Xeroderma pigmentosum (XP) A-G and V, Cockayne syndrome (CS) A and B, and trichothiodystrophy (TTD) group A, etc. This component, XPC, plays an important role in the early steps of global genome NER, especially in damage recognition, open complex formation, and repair protein complex formation. Mutations in this gene or some other NER components result in Xeroderma pigmentosum, a rare autosomal recessive disorder characterized by increased sensitivity to sunlight with the development of carcinomas at an early age. Alternatively spliced transcript variants have been found for this gene.
Specifications
| XPC | |
| Monoclonal | |
| 1.15 mg/mL | |
| PBS with no preservative | |
| P51612, Q01831 | |
| XPC | |
| Recombinant XPC purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.26 | |
| Unconjugated | |
| XPC | |
| DNA repair protein complementing XP-C cells; DNA repair protein complementing XP-C cells homolog; mutant xeroderma pigmentosum group C; NER; p125; RAD4; Xeroderma pigmentosum group C-complementing protein; Xeroderma pigmentosum group C-complementing protein homolog; xeroderma pigmentosum, complementation group C; XP3; XPC; XPC complex subunit, DNA damage recognition and repair factor; XPCC | |
| Mouse | |
| Protein G | |
| RUO | |
| 22591, 7508 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction