missing translation for 'onlineSavingsMsg'
Learn More

XPC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen XPC
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229338
View Documents
Novus Biologicals
NBP3-38529-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30230945
View Documents
Novus Biologicals
NBP3-38529-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

XPC Polyclonal antibody specifically detects XPC in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

XPC
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, DNA Repair, Nucleotide Excision Repair
PBS (pH 7.3), 50% glycerol
7508
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
complementation group C, p125, RAD4, Xeroderma pigmentosum group C-complementing protein, xeroderma pigmentosum, complementation group C, XP3, XPCCDNA repair protein complementing XP-C cells
Recombinant fusion protein containing a sequence corresponding to amino acids 700-940 of human XPC (NP_004619.3).,, Sequence:, GEVPYKMVKGFSNRARKARLAEPQLREENDLGLFGYWQTEEYQPPVAVDGKVPRNEFGNVYLFLPSMMPIGCVQLNLPNLHRVARKLDIDCVQAITGFDFHGGYSHPVTDGYIVCEEFKDVLLTAWENEQAVIERKEKEKKEKRALGNWKLLAKGLLIRERLKRRYGPKSEAAAPHTDAGGGLSSDEEEGTSSQAEAARILAASWPQNREDEEKQKLKGGPKKTKREKKAAASHLFPFEQL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.