missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ XPC Antibody (6D11), Novus Biologicals™

Product Code. 18345329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345329 Supplier Novus Biologicals™ Supplier No. H00007508M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XPC Monoclonal antibody specifically detects XPC in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPC
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 6D11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004619
Gene Alias complementation group C, p125, RAD4, Xeroderma pigmentosum group C-complementing protein, xeroderma pigmentosum, complementation group C, XP3, XPCCDNA repair protein complementing XP-C cells
Host Species Mouse
Immunogen XPC (NP_004619, 141 a.a. ∽ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EPVLGDVRESTAFSRSLLPVKPVEIEIETPEQAKTRERSEKIKLEFETYLRRAMKRFNKGVHEDTHKVHLLCLLANGFYRNNICSQPDLHAIGLSIIPARFTRVLPRDVD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 7508
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.