missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPB Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10102-100UL
This item is not returnable.
View return policy
Description
XPB Polyclonal specifically detects XPB in Human samples. It is validated for Western Blot.
Specifications
| XPB | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human XPB (NP_000113.1). Peptide sequence KRDRADRDKKKSRKRHYEDEEDDEEDAPGNDPQEAVPSAAGKQVDESGTK | |
| 100 μg | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2071 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction