missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576038
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne's syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
Specifications
| XPB | |
| Polyclonal | |
| Unconjugated | |
| Ercc3 | |
| Basic transcription factor 2 89 kDa subunit; BTF2; BTF2 p89; BTF2-p89; DNA excision repair protein ERCC-3; DNA repair helicase; DNA repair protein complementing XP-B cells; ERCC excision repair 3, TFIIH core complex helicase subunit; Ercc3; Ercc-3; excision repair 3; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; General transcription and DNA repair factor IIH helicase subunit XPB; GTF2H; RAD25; TFIIH; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p89; TFIIH subunit XPB; TTD2; Xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B; XPB; XPBC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13872, 2071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
| P19447, P49135 | |
| Ercc3 | |
| Recombinant full length Human XPB. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction