Learn More
Invitrogen™ XPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119824
Description
Positive Control: Hela cell lysate, A431 cell lysate, rat large intestine tissue, human thyroid tissue, human colon carcinoma tissue, human skin tissue, human prostate carcinoma tissue, human breast tissue, human breast carcinoma tissue, human stomach carcinoma tissue, human small intestine tissue, mouse testis tissue, mouse kidney tissue. Subcellular Location: Nucleus.
The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.
Specifications
| XPA | |
| Polyclonal | |
| Unconjugated | |
| XPA | |
| AI573865; DNA repair protein complementing XP-A cells; DNA repair protein complementing XP-A cells homolog; excision repair-controlling; mutant xeroderma pigmentosum complementation group A; xeroderma pigmentosum group A-complementing protein; Xeroderma pigmentosum group A-complementing protein homolog; xeroderma pigmentosum, complementation group A; XP1; XPA; XPA, DNA damage recognition and repair factor; Xpac | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22590, 298074, 7507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P23025, Q64267 | |
| XPA | |
| Synthetic peptide within human XPA aa 20-60. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.