missing translation for 'onlineSavingsMsg'
Learn More

XP32 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18348913
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348913 100 μg 100µL
18335125 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348913 Supplier Bio-Techne Supplier No. NBP317559100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XP32 Polyclonal antibody specifically detects XP32 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen XP32
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C1orf68, chromosome 1 open reading frame 68, LEP7, skin-specific protein (xp32), skin-specific protein 32, XP32
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: CQTYVKCPAPCQTTYVKCPTPCQTYVKCPAPCQMTYIKSPAPCQTQTCYVQGASPCQSYYVQAPASGSTSQYCVT
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100129271
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.