missing translation for 'onlineSavingsMsg'
Learn More

XMD8-92, MedChemExpress

Product Code. p-200101087
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486068 10 mg 10mg
16476466 100 mg 100mg
16472207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486068 Supplier Medchem Express Supplier No. HY14443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XMD8-92 is a potent ERK5 (BMK1)/BRD4 inhibitor with Kds of 80 and 190 nM, respectively. XMD8-92 inhibits DCAMKL2, PLK4 and TNK1 with Kds of 190, 600 and 890 nM, respectively. Anti-cancer activity.

XMD8-92 is a potent ERK5 (BMK1)/BRD4 inhibitor with Kds of 80 and 190 nM, respectively. XMD8-92 inhibits DCAMKL2, PLK4 and TNK1 with Kds of 190, 600 and 890 nM, respectively. Anti-cancer activity.

Specifications

Chemical Name or Material XMD8-92
CAS 1234480-50-2
Color White
Purity Grade Notes Research
Molecular Formula C26H30N6O3
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (105.36 mM; Need ultrasonic)
SMILES CN(C1=O)C2=CN=C(N=C2N(C3=C1C=CC=C3)C)NC4=C(C=C(C=C4)N5CCC(CC5)O)OCC
Molecular Weight (g/mol) 474.55
Formula Weight 474.55
Percent Purity 99.93%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.