missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ XMD 8-92

Description
ERK5 (BMK1) and BRD4 inhibitor (Kd values are 80 and 190 nM, respectively). Also inhibits DCAMKL2, PLK4 and TNK1 (Kd values are 190, 600 and 890 nM). Blocks growth factor-induced activation of cellular BMK1 and reduces BMK1 activity in in vitro kinase assays. Also reduces BMK1-dependent transactivating activity of MEF2C. Inhibits proliferation in a variety of cancer cell lines; blocks tumor cell proliferation and tumor-associated angiogenesis.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 474.55 |
| Purity | >98% |
| Molecular Weight (g/mol) | 474.565 |
| Chemical Name or Material | XMD 8-92 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?