Learn More
CELLPATH XMARK - TISSUE MARKING DYE
Description
- The optimised viscosity of the Xmark dye reduces viscosity on specimen application, achieving even dye coverage without dripping or running.
- The high pigment concentrations in XMark dye derive highly durable marking and easy application to diagnostic specimens.
- It coats the specimen tissue surface with minimal penetration. Its strong adhesion qualities mean the marking dye will not interfere with diagnostic interpretations, making them ideal for radical prostatectomies.
- Available in nine colours, all easy to distinguish, reducing potential errors and ensuring the colour shown on the bottle (macroscopic colours) matches the colour observed under the microscope. Unlike other dyes XMark is non-hazardous, negating the need for special requirements during use.
- Key Benefits:
- Emulsified pigments are blended perfectly with an aqueous base to provide a consistent and high-quality product every time
- Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both before and after tissue processing
- A 3-4 minute drying time improves laboratory workflow at the cut-up bench
- Non-flammable
Specifications
Specifications
| Capacity | 60 mL |
| Product Type | Tissue Marking Dye |
| Solution Type | Mordant Dye |
| Form | Liquid |