Learn More
Invitrogen™ xMAP Sheath Fluid Plus
Product Code. 17183935
Change view
Click to view available options
Quantity:
20 L
Unit Size:
Each
Product Code. Quantity unitSize
17183935 20 L Each
1 options
Product Code. 17183935 Supplier Invitrogen™ Supplier No. 4050021
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments.
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200 and FLEXMAP 3D systems.
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200 and FLEXMAP 3D systems.
This reformulated sheath product replaces the original xMAP Sheath Fluid (Cat. No. 4050015). The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
For Research Use Only. Not for use in diagnostic procedures. Not intended for human or animal therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20 L
Packaging Container