missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ XLT-4 Agar

Product Code. p-6448072
Click to view available options
Quantity:
500 g
Description:
XLT-4 Agar
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10585943

Brand: Thermo Scientific™ CM1061B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Efficiently isolate and identify salmonellae from clinical, environmental and food samples with highly selective Thermo Scientific™ XLT-4 Agar which reduces false-positives.

Efficiently isolate and identify salmonellae from clinical, environmental and food samples with highly selective Thermo Scientific™ XLT-4 Agar. XLT-4 Agar contains sodium tetradecylsulfate which acts to inhibit many of the background flora organisms whose growth can cause problems with differentiation on other salmonellae plating media. This enhanced selectivity allows a significant increase in the recovery of salmonellae, while reducing false-positive suspect colonies, saving time and reducing the costs of further identification of false-positive colonies1.

XLT-4 Agar is a highly selective plating medium used for isolation and identification of salmonellae from clinical, environmental and food samples according to Miller and Tate1. XLT-4 Agar has been shown to save time and reduce the costs of further identification of false presumptive positive colonies when compared to other salmonellae plating media1.
  • Ensures optimal growth of salmonellae while reducing false-positive suspect colonies
  • Inhibits unwanted background flora with Tergitol
  • Allows Salmonellae to be easily distinguished: appearing as black or red colonies with a black center 

The peptones and yeast extract in this formulation provide a source of amino nitrogen, along with essential nutrients and vitamins; this ensures optimal growth of salmonellae.

The selective agent is Tergitol 4 (also known as Niaproof 4 or sodium tetradecylsulfate), which is an anionic surfactant. This largely inhibits or reduces the growth of unwanted background flora. The background color of the plate is red, due to the inclusion of the phenol red, which will change color due to pH changes from fermentation and decarboxylation reactions.

Differentiation on this medium is facilitated by the fermentation of xylose, lactose and sucrose as well as the decarboxylation of lysine. Salmonellae appear as black or red colonies with a black center due to their ability to reduce thiosulphate to hydrogen sulphide, which causes the colony to blacken (as there are ferric ions present). Escherichia coli grow as yellow colonies as they are able to ferment the lactose in the formulation, causing a pH drop which shifts the color of the phenol indicator from red to yellow. Other organisms, such as shigellae, do not ferment the lactose, or reduce the thiosulphate, so appear as clear red colonies with no blackening.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.


TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 10°C to 30°C
Description XLT-4 Agar
Form Powder
Manufacturing Origin United Kingdom
Product Type Agar
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.