Learn More
Invitrogen™ XLF Recombinant Rabbit Monoclonal Antibody (24GB480)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557513
 View more versions of this product
Product Code. 30276268
  • £172.00 / 20µL
Description
XLF Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
XLF
Recombinant Monoclonal
Unconjugated
NHEJ1
1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor
Rabbit
Affinity chromatography
RUO
79840
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB480
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H9Q4
NHEJ1
A synthesized peptide derived from human XLF (aa 150-299).
20 μL