missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XLF Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518584
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
Specifications
| XLF | |
| Polyclonal | |
| Unconjugated | |
| NHEJ1 | |
| 1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 79840 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H9Q4 | |
| NHEJ1 | |
| Synthetic peptide sequence (QRPQLSKVKRKKPR) corresponding to the C-terminus amino acids of NHEJ1 (aa 282-296). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction