Learn More
Invitrogen™ XLF Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA517198
Product Code. 11516021
  • £392.00 / 100µL
Description
It is not recommended to aliquot this antibody.
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
XLF
Polyclonal
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Q9H9Q4
NHEJ1
Synthetic peptide corresponding to amino acids near the carboxy terminus of human XLF.
100 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Western Blot
Unconjugated
NHEJ1
1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor
Rabbit
Antigen Affinity Chromatography
RUO
79840
-20°C