Learn More
XLF Mouse, anti-Human, CoraLite Plus 750, Clone: 1F8C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL750-66552-100UL
Description
The XLF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human XLF. This antibody recognizes human antigen. The XLF antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.
Specifications
| XLF | |
| 1F8C7 | |
| CoraLite Plus 750 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2b |
| Western Blot | |
| 1000 μg/mL | |
| FLJ12610, NHEJ1, Protein cernunnos, XLF, XRCC4 like factor | |
| 100 μL | |
| 79840 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |