missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XLF Antibody (S08-9D7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19804-100ul
This item is not returnable.
View return policy
Description
XLF Monoclonal antibody specifically detects XLF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| XLF | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-9D7 | |
| Unconjugated | |
| Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor | |
| Recombinant protein of human XLF (Uniprot # Q9H9Q4) | |
| 100 μg | |
| Cancer, Chromatin Research, DNA Repair | |
| 79840 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction