missing translation for 'onlineSavingsMsg'
Learn More

XL888, MedChemExpress

Product Code. 16465638
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465638 10 mg 10mg
16461339 1 mg 1mg
16451807 50 mg 50mg
16443356 25 mg 25mg
16428337 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16465638 Supplier Medchem Express Supplier No. HY13313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XL888 is a heat shock protein-90 (HSP90) inhibitor, with an IC50 of 24 nM.

XL888 is a heat shock protein-90 (HSP90) inhibitor, with an IC50 of 24 nM.

Specifications

Chemical Name or Material XL888
CAS 1149705-71-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H37N5O3
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (99.28 mM; Need ultrasonic)
SMILES CC1=CC(C(N)=O)=C(N[C@H](C)CC)C=C1C(N[C@]2([H])C[C@H](CC3)N(C4=NC=C(C(C5CC5)=O)C=C4)[C@H]3C2)=O
Molecular Weight (g/mol) 503.64
Formula Weight 503.64
Percent Purity 98.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.