missing translation for 'onlineSavingsMsg'
Learn More

XL413, TRC

Product Code. 30454984
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454984 25 mg 25mg
30452405 5 mg -
30449097 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454984 Supplier Toronto Research Chemicals Supplier No. TRCX74623025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1169562-71-3
Molecular Formula C14H13Cl2N3O2
Molecular Weight (g/mol) 326.18
Synonym 8-Chloro-2-(2S)-2-pyrrolidinyl-benzofuro[3,2-d]pyrimidin-4(3H)-one Hydrochloride, BMS-863233
IUPAC Name 8-chloro-2-[(2S)-pyrrolidin-2-yl]-3H-[1]benzofuro[3,2-d]pyrimidin-4-one;hydrochloride
SMILES n1ccc(cc1)CCSCCc2ccncc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H12ClN3O2.ClH/c15-7-3-4-10-8(6-7)11-12(20-10)14(19)18-13(17-11)9-2-1-5-16-9;/h3-4,6,9,16H,1-2,5H2,(H,17,18,19);1H/t9-;/m0./s1
Formula Weight 325.0385
Chemical Name or Material XL413
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.